L-Isoleucine methyl ester hydrochloride, 98+%, Thermo Scientific Chemicals
L-Isoleucine methyl ester hydrochloride, 98+%, Thermo Scientific Chemicals
L-Isoleucine methyl ester hydrochloride, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

L-Isoleucine methyl ester hydrochloride, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
A17847.03
also known as A17847-03
1g
A17847.06
also known as A17847-06
5g
A17847.14
also known as A17847-14
25g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number A17847.03
also known as A17847-03
Price (USD)
-
Quantity:
1g
Request bulk or custom format
Chemical Identifiers
CAS18598-74-8
IUPAC Namemethyl (2S,3S)-2-amino-3-methylpentanoate hydrochloride
Molecular FormulaC7H16ClNO2
InChI KeyGGTBEWGOPAFTTH-GEMLJDPKSA-N
SMILESCl.CC[C@H](C)[C@H](N)C(=O)OC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream
FormCrystals or powder or crystalline powder
Assay (Titration ex Chloride)≥98.0 to ≤102.0% (non-U.S. specification)
Assay (HPLC)≥98.0% (U.S. specification)
CommentSpecification differs for U.S. and non-U.S. material where indicated
View more
L-Isoleucine methyl ester used for chemical synthesis. E.g. It can also be used to produce 3-methyl-2-(4-nitro-imidazol-1-yl)-pentanoic acid methyl ester. It also finds its use as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Isoleucine methyl ester used for chemical synthesis. E.g. It can also be used to produce 3-methyl-2-(4-nitro-imidazol-1-yl)-pentanoic acid methyl ester. It also finds its use as a pharmaceutical intermediate.

Solubility
Soluble in methanol (50 mg/ml clear, colorless solution).

Notes
Hygroscopic. Keep away from oxidizing agents, water/moisture. Store under inert atmosphere. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. In order to obtain a higher solubility, warm the tube at 37°C and shake it in the ultrasonic bath for a while.
RUO – Research Use Only
I B Rozenval’d. [The stimulation of the transition of resting NIH 3T3 cells to proliferation under exposure to lysosomal inhibitors].Tsitologiia.,1990,32(3), 220-226.

S C Alfieri.; V Zilberfarb.; M Rabinovitch. Destruction of Leishmania mexicana amazonensis amastigotes by leucine methyl ester: protection by other amino acid esters.Parasitology.;1987,95(Pt 1), 31-41.