Thermo Scientific Chemicals

Tin(II) oxide, 99%, Thermo Scientific Chemicals

Catalog number: A17889.22
100 g, Each
Thermo Scientific Chemicals

Tin(II) oxide, 99%, Thermo Scientific Chemicals

Catalog number: A17889.22
100 g, Each
Quantity
Catalog number: A17889.22
also known as A17889-22
Price (USD)
Price: 45.80
Online price: 38.65
Your price:
Quantity
-

Chemical Identifiers

CAS
21651-19-4
Assay from Supplier's CofA
Tin (II): 87.0 to 87.5%
Elemental Analysis
Lead, Pb ≤ 150ppm
Water Content (Karl Fischer Titration)
≤0.1%
Appearance (Color)
Dark grey to blue-black
Elemental Analysis
Iron, Fe ≤ 60ppm

Description

Tin(II) oxide is used as reducing agent, soft abrasive, and in preparation of stannous salts. It is used in the manufacture of copper ruby glass, and for illumination with UV light.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tin(II) oxide is used as reducing agent, soft abrasive, and in preparation of stannous salts. It is used in the manufacture of copper ruby glass, and for illumination with UV light.

Solubility
Insoluble in water and alcohol.

Notes
Incompatible with materials like strong oxidizing agents, strong acids, strong reducing agents and alkali metals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text