Triethyl phosphate, 98+%
Triethyl phosphate, 98+%
Triethyl phosphate, 98+%
Triethyl phosphate, 98+%
Thermo Scientific Chemicals

Triethyl phosphate, 98+%

CAS: 78-40-0 | C6H15O4P | 182.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2500 g
Catalog number A17938.36
also known as A17938-36
Price (TWD)
-
Request A Quote
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
CAS78-40-0
IUPAC Nametriethyl phosphate
Molecular FormulaC6H15O4P
InChI KeyDQWPFSLDHJDLRL-UHFFFAOYSA-N
SMILESCCOP(=O)(OCC)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥98.0% (non-U.S. specification)
Assay from Suppliers CofA≥98.0% (U.S. specification)
CommentSpecification differs for U.S. and non-U.S. material where indicated
View more

Triethyl phosphate finds it major applications in plastics industry as a flame retardant, plasticizer and carrier, where it is available in the matrix. A further 10 to 20 % are used in other industrial branches as a solvent, plasticizer, flame retardant or intermediate for the production of pharmaceuticals, and lacquers. Triethyl phosphate is a useful synthetic intermediate used in the synthesis of mesoporous spheres of metal oxides and phosphates. It is also used as an industrial catalyst employed in ketene synthesis where the compound is hydrolyzed, as a polymer resin modifier, as a solvent, a car paint repairing product and as flame retarder

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triethyl phosphate finds it major applications in plastics industry as a flame retardant, plasticizer and carrier, where it is available in the matrix. A further 10 to 20 % are used in other industrial branches as a solvent, plasticizer, flame retardant or intermediate for the production of pharmaceuticals, and lacquers. Triethyl phosphate is a useful synthetic intermediate used in the synthesis of mesoporous spheres of metal oxides and phosphates. It is also used as an industrial catalyst employed in ketene synthesis where the compound is hydrolyzed, as a polymer resin modifier, as a solvent, a car paint repairing product and as flame retarder

Solubility
Fully miscible in water at 20°C.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

General References:

  1. K Troev.; G Grancharov.; R Tsevi.; A Tsekova. A novel approach to recycling of polyurethanes: chemical degradation of flexible polyurethane foams by triethyl phosphate.Polymer.;2000,41(19), 7017-7022.
  2. Ronald V. Hodges.; T. J. McDonnell.; J. L. Beauchamp. Properties and reactions of trimethyl phosphite, trimethyl phosphate, triethyl phosphate, and trimethyl phosphorothionate by ion cyclotron resonance spectroscopy.J. Am. Chem. Soc.,1980,102(4), 1327-1332.