Methyl 4-methoxyphenylacetate, 97+%
Methyl 4-methoxyphenylacetate, 97+%
Methyl 4-methoxyphenylacetate, 97+%
Methyl 4-methoxyphenylacetate, 97+%
Thermo Scientific Chemicals

Methyl 4-methoxyphenylacetate, 97+%

CAS: 23786-14-3 | C10H12O3 | 180.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number A17940.14
also known as A17940-14
Price (USD)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS23786-14-3
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Refractive Index1.5135-1.5185 @ 20?C
Assay (GC)≥97.0%
Appearance (Color)Clear colorless to pale yellow
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only