Hydroxypropyl methacrylate, mixture of isomers, 97+%, stab. with ca 0.02% 4-methoxyphenol
Hydroxypropyl methacrylate, mixture of isomers, 97+%, stab. with ca 0.02% 4-methoxyphenol
Hydroxypropyl methacrylate, mixture of isomers, 97+%, stab. with ca 0.02% 4-methoxyphenol
Thermo Scientific Chemicals

Hydroxypropyl methacrylate, mixture of isomers, 97+%, stab. with ca 0.02% 4-methoxyphenol

CAS: 27813-02-1 | C7H12O3 | 144.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1000 mL
5000 mL
Catalog number A17951.AK
also known as A17951-AK
Price (EUR)
35,60
Each
Quantity:
250 mL
Request bulk or custom format
Price (EUR)
35,60
Each
Chemical Identifiers
CAS27813-02-1
IUPAC Name1-hydroxypropan-2-yl 2-methylprop-2-enoate
Molecular FormulaC7H12O3
InChI KeyZMARGGQEAJXRFP-UHFFFAOYNA-N
SMILESCC(CO)OC(=O)C(C)=C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay (GC)≥97.0% (mixture of isomers)
Stabilizer150 tO 300 MEHQ
CommentA mixture of hydroxypropyl and hydroxyisopropyl methacrylates
Refractive Index1.4460-1.4520 @ 20°C
View more
Hydroxypropyl methacrylate is a functional monomer for the preparation of hot solid acrylic coatings, styrene-butadiene latex modifiers, acrylic modified polyurethane coating ,water-soluble plating coatings , adhesives ,textile finishing agent ,paper coating , photosensitive paint and polyurethane vinyl resin modified agent. Mainly used in the manufacture are active groups of hydroxyl acrylic resin. Used in reactive diluent and cross-linking agent in the UV curing system.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hydroxypropyl methacrylate is a functional monomer for the preparation of hot solid acrylic coatings, styrene-butadiene latex modifiers, acrylic modified polyurethane coating ,water-soluble plating coatings , adhesives ,textile finishing agent ,paper coating , photosensitive paint and polyurethane vinyl resin modified agent. Mainly used in the manufacture are active groups of hydroxyl acrylic resin. Used in reactive diluent and cross-linking agent in the UV curing system.

Solubility
Soluble in water.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only

General References:

  1. Elizabeth H. Leduc and S. J. Holt. Hydroxypropyl Methacrylate, A New Water-Miscible Embedding Medium For Electron Microscopy. JCB Home. 1965, 26, (1), 137-155.
  2. Murakami T; Itoshima T; Hitomi K; Ohtsuka A; Jones AL. A monomeric methyl and hydroxypropyl methacrylate injection medium and its utility in casting blood capillaries and liver bile canaliculi for scanning electron microscopy.. Arch Histol Jpn. 1984, 47, (2), 223-237.