Thermo Scientific™
Document Holder Kit
Related applications:
Anatomical Pathology
The Thermo Scientific™ Document Holder Kit is designed to work in conjunction with the Thermo Scientific™ HistoStar™ Embedding Workstation. It fits to the top of the embedding station and allows documents to be viewed during operation.
Back to top
Description
The Thermo Scientific™ Document Holder Kit is designed to work in conjunction with the Thermo Scientific™ HistoStar™ Embedding Workstation. It fits to the top of the embedding station and allows documents to be viewed during operation.