Thermo Scientific Chemicals

Magnesium carbonate hydroxide pentahydrate, light, 98%, Thermo Scientific Chemicals

Catalog number: A18070.0I
5000 g, Each
Thermo Scientific Chemicals

Magnesium carbonate hydroxide pentahydrate, light, 98%, Thermo Scientific Chemicals

Catalog number: A18070.0I
5000 g, Each
Quantity
Catalog number: A18070.0I
also known as A18070-0I
Price (USD)
Price: 277.00
Online price: 239.65
Your price:
Quantity
-

Chemical Identifiers

CAS
56378-72-4
IUPAC Name
pentamagnesium(2+) pentahydrate dihydroxide tetracarbonate
Molecular Formula
C4H12Mg5O19
InChI Key
SXKPPTNZIUYFJR-UHFFFAOYSA-D
SMILES
O.O.O.O.O.[OH-].[OH-].[Mg++].[Mg++].[Mg++].[Mg++].[Mg++].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O
Complexometric Titration
≥97.5 to ≤102.5%
Identification (FTIR)
Conforms
Appearance (Color)
White
Form
Powder

Description

Magnesium carbonate hydroxide pentahydrate is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Magnesium carbonate hydroxide pentahydrate is used as pharmaceutical intermediate.

Solubility
Soluble in water and acids. Insoluble in alcohol.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text