Thermo Scientific Chemicals

4-Methyl-1H-pyrazole, 97+%, Thermo Scientific Chemicals

Catalog number: A18083.06
5 g, Each
Thermo Scientific Chemicals

4-Methyl-1H-pyrazole, 97+%, Thermo Scientific Chemicals

Catalog number: A18083.06
5 g, Each
Quantity
Catalog number: A18083.06
also known as A18083-06
Price (USD)
Price: 321.00
Online price: 277.65
Your price:
Quantity
-

Chemical Identifiers

CAS
7554-65-6
IUPAC Name
4-methyl-1H-pyrazole
Molecular Formula
C4H6N2
InChI Key
RIKMMFOAQPJVMX-UHFFFAOYSA-N
SMILES
CC1=CNN=C1
Appearance (Color)
Clear colorless to yellow
Assay (GC)
≥97.0%
Form
Liquid
Refractive Index
1.4915-1.4965 @ 20?C

Description

4-Methyl-1H-pyrazole is used as an antidote in methanol and ethylene glycol poisoning. It is also employed as an alcohol dehydrogenase inhibitor. It acts a ligand in coordination chemistry. Further, it serves as an intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methyl-1H-pyrazole is used as an antidote in methanol and ethylene glycol poisoning. It is also employed as an alcohol dehydrogenase inhibitor. It acts a ligand in coordination chemistry. Further, it serves as an intermediate in organic synthesis.

Solubility
Miscible with water, ether, benzene and alcohol.

Notes
Hygroscopic. Incompatible withstrong oxidizing agents, strong acids, acid chlorides, acid anhydrides and chloroformates.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text