4-(2-Pyridylazo)resorcinol monosodium salt monohydrate
4-(2-Pyridylazo)resorcinol monosodium salt monohydrate
4-(2-Pyridylazo)resorcinol monosodium salt monohydrate
Thermo Scientific Chemicals

4-(2-Pyridylazo)resorcinol monosodium salt monohydrate

CAS: 16593-81-0 | C11H9N3O2 | 215.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number A18103.14
also known as A18103-14
Price (USD)
306.65
Special offer
Online exclusive
Ends: 30-Jun-2026
360.00
Save 53.35 (15%)
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
306.65
Special offer
Online exclusive
Ends: 30-Jun-2026
360.00
Save 53.35 (15%)
Each
Chemical Identifiers
CAS16593-81-0
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Orange to brown
FormCrystals or powder or crystalline powder
Specific Absorbance (E 1%/1cm)min. 1100 (at 413nm in water)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only