Thermo Scientific Chemicals

2-Chlorothioxanthone, 99%, Thermo Scientific Chemicals

25g, Each
Thermo Scientific Chemicals

2-Chlorothioxanthone, 99%, Thermo Scientific Chemicals

25g, Each
Quantity
Catalog number: A18131.14
also known as A18131-14
Price (USD)
Price: 64.70
Your Price:
Quantity
-

Chemical Identifiers

CAS
86-39-5
Form
Powder
Assay (GC)
≥98.5%
Melting Point (clear melt)
145.5-151.5?C
Appearance (Color)
Yellow to pale brown

Description

2-Chlorothioxanthone is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chlorothioxanthone is used as pharmaceutical intermediate.

Solubility
Partly miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only