6-Nitropiperonyl alcohol, 98+%
6-Nitropiperonyl alcohol, 98+%
6-Nitropiperonyl alcohol, 98+%
Thermo Scientific Chemicals

6-Nitropiperonyl alcohol, 98+%

CAS: 15341-08-9 | C8H7NO5 | 197.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A18140.06
also known as A18140-06
Price (EUR)
49,40
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
49,40
Each
Chemical Identifiers
CAS15341-08-9
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥98.0%
FormPowder
Melting Point (clear melt)120.5-126.5?C
Appearance (Color)Yellow
6-Nitropiperonyl alcohol is used as an organic chemical synthesis intermediate. It is also used as a biochemical for proteomics research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Nitropiperonyl alcohol is used as an organic chemical synthesis intermediate. It is also used as a biochemical for proteomics research.

Notes
Light sensitive. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. D.V.Carter; P.T.Charlton; A.H.Fenton; J.R.Housley; B.Lessel. The preparation and the antibacterial and antifungal properties of some substituted benzyl alcohols. Journal of Pharmacy and Pharmacology. 1958, 10,(S1), 149T-159T.
  2. J.Meuche-Albeny; P.Rathelot; M.P.Crozet; P.Vanelle. S RN1 reactions in the nitrobenzo[1,3]dioxole series. Chemistry of Heterocyclic Compounds. 2003, 39,(8), 989-997.