Thermo Scientific Chemicals

6-Nitropiperonyl alcohol, 98+%, Thermo Scientific Chemicals

Catalog number: A18140.14
25 g, Each
Thermo Scientific Chemicals

6-Nitropiperonyl alcohol, 98+%, Thermo Scientific Chemicals

Catalog number: A18140.14
25 g, Each
Quantity
Catalog number: A18140.14
also known as A18140-14
Price (USD)
Price: 529.00
Online price: 461.65
Your price:
Quantity
-

Chemical Identifiers

CAS
15341-08-9
Form
Powder
Appearance (Color)
Yellow
Melting Point (clear melt)
120.5-126.5?C
Assay (GC)
≥98.0%

Description

6-Nitropiperonyl alcohol is used as an organic chemical synthesis intermediate. It is also used as a biochemical for proteomics research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Nitropiperonyl alcohol is used as an organic chemical synthesis intermediate. It is also used as a biochemical for proteomics research.

Notes
Light sensitive. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text