Methylene Blue, high purity, biological stain
Methylene Blue, high purity, biological stain
Methylene Blue, high purity, biological stain
Thermo Scientific Chemicals

Methylene Blue, high purity, biological stain

CAS: 122965-43-9 | C16H18ClN3S | 319.85 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number A18174.22
also known as A18174-22
Price (EUR)
65,80
Each
Quantity:
100 g
Request bulk or custom format
Price (EUR)
65,80
Each
Chemical Identifiers
CAS122965-43-9
IUPAC Name7-(dimethylamino)-N,N-dimethyl-3H-phenothiazin-3-iminium chloride
Molecular FormulaC16H18ClN3S
InChI KeyCXKWCBBOMKCUKX-UHFFFAOYSA-M
SMILES[Cl-].CN(C)C1=CC=C2N=C3C=CC(C=C3SC2=C1)=[N+](C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
Loss on Drying≤18% (105?C/constant weight)
lambda-max (UV-Vis)662-666nm (water)
Appearance (Color)Dark green
FormCrystals or powder or crystalline powder
View more
Methylene blue is used as a dye in different staining procedures viz. Wright's stain and Jenner's stain. It also serves as an indicator and medicine. Further, it is used to examine RNA and DNA under the microscope or in a gel. It is widely used as a redox indicator in analytical chemistry. It acts as a photosensitizer utilized to generate singlet oxygen in order to make organic peroxides by a Diels-Alder reaction. It is also used in the sulfide analysis. It plays an important role in aquaculture to treat fungal infections.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methylene blue is used as a dye in different staining procedures viz. Wright′s stain and Jenner′s stain. It also serves as an indicator and medicine. Further, it is used to examine RNA and DNA under the microscope or in a gel. It is widely used as a redox indicator in analytical chemistry. It acts as a photosensitizer utilized to generate singlet oxygen in order to make organic peroxides by a Diels-Alder reaction. It is also used in the sulfide analysis. It plays an important role in aquaculture to treat fungal infections.

Solubility
Soluble in water, alcohol, acetic acid, glycerol and chloroform. Insoluble in ether.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Chowdhury, P. R.; Bhattacharyya, K. G. Ni/Ti layered double hydroxide: synthesis, characterization and application as a photocatalyst for visible light degradation of aqueous methylene blue. Dalton Trans. 2015, 44 (15), 6809-6824.
  2. Das, M.; Bhattacharyya, K. G. Use of Raw and Acid-Treated MnO2 as Catalysts for Oxidation of Dyes in Water: A Case Study with Aqueous Methylene Blue. Chem. Eng. Commun. 2014, 202 (12), 1657-1667.