Thermo Scientific Chemicals

Sodium hydrogen arsenate heptahydrate, 98%, Thermo Scientific Chemicals

Catalog number: A18275.22
100 g, Each
Thermo Scientific Chemicals

Sodium hydrogen arsenate heptahydrate, 98%, Thermo Scientific Chemicals

Catalog number: A18275.22
100 g, Each
Quantity
Catalog number: A18275.22
also known as A18275-22
Price (USD)

Chemical Identifiers

CAS
10048-95-0
IUPAC Name
disodium heptahydrate hydrogen arsorate
Molecular Formula
AsH15Na2O11
InChI Key
KOLXPEJIBITWIQ-UHFFFAOYSA-L
SMILES
O.O.O.O.O.O.O.[Na+].[Na+].O[As]([O-])([O-])=O
Impurity content
Iron, Fe: ≤0.001%
Appearance (Color)
White
Form
Crystals or powder or crystalline powder
Impurity content
Arsenite, AsO2: ≤0.01%
Impurity content
Nitrate, NO3: ≤0.005%

Description

Sodium hydrogen arsenate heptahydrate is used as a source of soluble arsenic and in dyeing and printing with Turkey-red oil. It also acts as a veterinary drug for anti-parasitic, skin diseases, blood diseases, rheumatism, and asthma.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water and glycerol.

Notes
Incompatible with strong oxidizing agents and strong acids.
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text