Glutaconic aldehyde dianilide hydrochloride, mixture of isomers, 98%
Glutaconic aldehyde dianilide hydrochloride, mixture of isomers, 98%
Glutaconic aldehyde dianilide hydrochloride, mixture of isomers, 98%
Thermo Scientific Chemicals

Glutaconic aldehyde dianilide hydrochloride, mixture of isomers, 98%

CAS: 1497-49-0 | C17H17ClN2 | 284.79 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A18390.22
also known as A18390-22
Price (TWD)
-
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS1497-49-0
SpecificationsSpecification SheetSpecification Sheet
Assay (Titration ex Chloride)≥97.5 to ≤102.5%
Appearance (Color)Purple to red to dark red
Melting Point (clear melt)125.0-132.0?C
FormPowder
Glutaconic aldehyde dianilide hydrochloride is a tricarbocyanine laser dye intermediate. It is used as an intermediate for pharmaceuticals and dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Glutaconic aldehyde dianilide hydrochloride is a tricarbocyanine laser dye intermediate. It is used as an intermediate for pharmaceuticals and dyes.

Solubility
Soluble in methanol. Insoluble in water..

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is hygroscopic in nature.
RUO – Research Use Only

General References:

  1. Kai Licha.; Sarah Bhargava.; Christiane Rheinländer.; Andreas Becker.; Jens Schneider-Mergener.; Rudolf Volkmer-Engert. Highly parallel nano-synthesis of cleavable peptide-dye conjugates on cellulose membranes. Tetrahedron Letters. 2000, 41 (11), 1711-1715.
  2. Zongren Zhang.; Kexian Liang.; Sharon Bloch.; Mikhail Berezin.; Samuel Achilefu. Monomolecular Multimodal Fluorescence-Radioisotope Imaging Agents. Bioconjugate Chem. 2005, 16 (5), 1232-1239.
  3. Stable source of glutaconic aldehyde.