7-Hydroxy-5-methyl[1,2,4]triazolo[1,5-a]pyrimidine, 98%
7-Hydroxy-5-methyl[1,2,4]triazolo[1,5-a]pyrimidine, 98%
7-Hydroxy-5-methyl[1,2,4]triazolo[1,5-a]pyrimidine, 98%
7-Hydroxy-5-methyl[1,2,4]triazolo[1,5-a]pyrimidine, 98%
Thermo Scientific Chemicals

7-Hydroxy-5-methyl[1,2,4]triazolo[1,5-a]pyrimidine, 98%

CAS: 2503-56-2 | C6H6N4O | 150.141 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number A18438.14
also known as A18438-14
Price (EUR)
45,05
Online Exclusive
53,00
Save 7,95 (15%)
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (EUR)
45,05
Online Exclusive
53,00
Save 7,95 (15%)
Each
Add to cart
Chemical Identifiers
CAS2503-56-2
IUPAC Name5-methyl-1H,7H-[1,2,4]triazolo[1,5-a]pyrimidin-7-one
Molecular FormulaC6H6N4O
InChI KeyINVVMIXYILXINW-UHFFFAOYSA-N
SMILESCC1=CC(=O)N2NC=NC2=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Melting Point (clear melt)282.0-288.0?C
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
 
RUO – Research Use Only