D-Panthenol, 98+%
D-Panthenol, 98+%
D-Panthenol, 98+%
Thermo Scientific Chemicals

D-Panthenol, 98+%

CAS: 81-13-0 | C9H19NO4 | 205.25 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
100 g
500 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
產品號碼 A18499.22
亦稱為 A18499-22
價格 (TWD)
-
Quantity:
100 g
Request bulk or custom format
化學識別
CAS81-13-0
IUPAC Name(2R)-2,4-dihydroxy-N-(3-hydroxypropyl)-3,3-dimethylbutanamide
Molecular FormulaC9H19NO4
InChI KeySNPLKNRPJHDVJA-ZETCQYMHSA-N
SMILESCC(C)(CO)[C@@H](O)C(=O)NCCCO
檢視更多
規格Specification Sheet規格表
Appearance (Color)Clear colorless
Water Content (Karl Fischer Titration)≤1.0%
FormViscous liquid
Identification (FTIR)Conforms
Assay from Supplier's CofA≥98.0 to ≤102.0%
檢視更多
D-Panthenol is used as precursors in the biosynthesis of coenzyme A. D-Panthenol and D-Pantothenic acid are used in a variety of skin protection products. D-pantothenic acid have a role in controlling keratinocyte proliferation and differentiation. D-Panthenol is used for studies of skin protection emulsions from UV irradiation and as a humectant. It is also used in creams, lotions, shampoos, conditioners and body washes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-Panthenol is used as precursors in the biosynthesis of coenzyme A. D-Panthenol and D-Pantothenic acid are used in a variety of skin protection products. D-pantothenic acid have a role in controlling keratinocyte proliferation and differentiation. D-Panthenol is used for studies of skin protection emulsions from UV irradiation and as a humectant. It is also used in creams, lotions, shampoos, conditioners and body washes.

Solubility
Fully miscible in water and soluble in alcohol.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is hygroscopic in nature.
RUO – Research Use Only

General References:

  1. Lai-Hao Wang.; Shih-Wen Tseng. Direct determination of d-panthenol and salt of pantothenic acid in cosmetic and pharmaceutical preparations by differential pulse voltammetry. Analytica Chimica Acta. 2001, 432 (1), 39-48.
  2. Rajeshwari V. Hosahalli.; Anita P. Savanur.; Sharanappa T. Nandibewoor.; Shivamurti A. Chimatadar. Kinetics and mechanism of uncatalysed and ruthenium(III)-catalysed oxidation of d-panthenol by alkaline permanganate. Transition Metal Chemistry. 2010, 35 (2), 237-246.