2'-Hydroxy-6'-methoxyacetophenone, 97%
2'-Hydroxy-6'-methoxyacetophenone, 97%
2'-Hydroxy-6'-methoxyacetophenone, 97%
Thermo Scientific Chemicals

2'-Hydroxy-6'-methoxyacetophenone, 97%

CAS: 703-23-1 | C9H10O3 | 166.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number A18539.03
also known as A18539-03
Price (EUR)
44,90
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
44,90
Each
Chemical Identifiers
CAS703-23-1
IUPAC Name1-(2-hydroxy-6-methoxyphenyl)ethan-1-one
Molecular FormulaC9H10O3
InChI KeyUENLHUMCIOWYQN-UHFFFAOYSA-N
SMILESCOC1=CC=CC(O)=C1C(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Melting Point (clear melt)55.0-61.0?C
Appearance (Color)Yellow
Assay (GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only