4-Phenylazodiphenylamine, 96%
4-Phenylazodiphenylamine, 96%
4-Phenylazodiphenylamine, 96%
Thermo Scientific Chemicals

4-Phenylazodiphenylamine, 96%

CAS: 101-75-7 | C18H15N3 | 273.34 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A18612.14
also known as A18612-14
Price (USD)
-
Quantity:
25 g
Request bulk or custom format
Chemical Identifiers
CAS101-75-7
IUPAC NameN-phenyl-4-(2-phenyldiazen-1-yl)aniline
Molecular FormulaC18H15N3
InChI KeyVXLFYNFOITWQPM-UHFFFAOYSA-N
SMILESN(C1=CC=CC=C1)C1=CC=C(C=C1)N=NC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Green to green-brown to brown
Assay (Non-aqueous acid-base Titration)≥95.0 to ≤105.0%
Assay (HPLC)≥95.0%
Melting Point (clear melt)78.0-87.0°C
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only