Ácido 5-cloroindol-2-carboxílico, 98 %, Thermo Scientific Chemicals
Ácido 5-cloroindol-2-carboxílico, 98 %, Thermo Scientific Chemicals
Ácido 5-cloroindol-2-carboxílico, 98 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ácido 5-cloroindol-2-carboxílico, 98 %, Thermo Scientific Chemicals

CAS: 10517-21-2 | C9H5ClNO2 | 194.59 g/mol
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
1 g
5 g
25 g
Número de catálogo A18626.03
también denominado A18626-03
Precio (USD)
-
Cantidad:
1 g
Pedido a granel o personalizado
Especificaciones
Beilstein153229
CAS10517-21-2
Nombre del producto químico o material5-Chloroindole-2-carboxylic acid
Número EINECS234-050-1
Formula Weight (peso de la fórmula)195.6
Ver más
Potent antagonist at the glycine site of the NMDA receptor

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only