Thermo Scientific Chemicals

DL-2-Hydroxybutyric acid sodium salt, 97+%, Thermo Scientific Chemicals

製品番号(カタログ番号): A18636.14
25 g, Each
Thermo Scientific Chemicals

DL-2-Hydroxybutyric acid sodium salt, 97+%, Thermo Scientific Chemicals

製品番号(カタログ番号): A18636.14
25 g, Each
数量
製品番号(カタログ番号): A18636.14
または、製品番号A18636-14

化学物質識別子

CAS
5094-24-6
IUPAC Name
sodium 2-hydroxybutanoate
Molecular Formula
C4H7NaO3
InChI Key
MOSCXNXKSOHVSQ-UHFFFAOYNA-M
SMILES
[Na+].CCC(O)C([O-])=O
Appearance (Color)
White
Assay (Non-aqueous acid-base Titration)
≥97.0 to ≤103.0%
Form
Crystals or powder or crystalline powder
Water Content (Karl Fischer Titration)
≤3.0%
Identification (FTIR)
Conforms

概要

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

ドキュメントおよびダウンロード

証明書

    よくあるご質問(FAQ)

    引用および参考文献

    Search citations by name, author, journal title or abstract text