Thermo Scientific Chemicals

Acetylenedicarboxylic acid, 97%, Thermo Scientific Chemicals

Catalog number: A18736.22
100 g, Each
Thermo Scientific Chemicals

Acetylenedicarboxylic acid, 97%, Thermo Scientific Chemicals

Catalog number: A18736.22
100 g, Each
Quantity
Catalog number: A18736.22
Price (USD)
Price: 325.00
Online price: 276.65
Your price:
Quantity
-

Chemical Identifiers

CAS
142-45-0
IUPAC Name
but-2-ynedioic acid
Molecular Formula
C4H2O4
InChI Key
YTIVTFGABIZHHX-UHFFFAOYSA-N
SMILES
OC(=O)C#CC(O)=O
Appearance (Color)
White to cream to yellow to pale brown
Identification (FTIR)
Conforms
Form
Powder or crystalline powder or crystals
Assay (HPLC)
≥96.0%
Water Content (Karl Fischer Titration)
≤1.5%

Description

Acetylenedicarboxylic acid is employed in the preparation of dimethyl acetylenedicarboxylate and potassium hydrogenacetylenedicarboxylate. It is also used as a laboratory reagent. Further, it is used as a precursor in organic synthesis and also serves as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Acetylenedicarboxylic acid is employed in the preparation of dimethyl acetylenedicarboxylate and potassium hydrogenacetylenedicarboxylate. It is also used as a laboratory reagent. Further, it is used as a precursor in organic synthesis and also serves as a pharmaceutical intermediate.

Solubility
Soluble in diethyl ether and water.

Notes
Air sensitive. Store in a cool place. Incompatible with bases, alkali, reducing agents and oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text