2,5-Difluorophenol, 97%
2,5-Difluorophenol, 97%
2,5-Difluorophenol, 97%
2,5-Difluorophenol, 97%
Thermo Scientific Chemicals

2,5-Difluorophenol, 97%

CAS: 2713-31-7 | C6H4F2O | 130.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A18744.14
also known as A18744-14
Price (EUR)
115,00
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (EUR)
115,00
Each
Add to cart
Chemical Identifiers
CAS2713-31-7
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Melting Point (clear melt)32.0-42.0?C
FormCrystals or powder or crystalline powder or fused solid
Assay (GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only