Thermo Scientific Chemicals

Dimethyl maleate, 96%, Thermo Scientific Chemicals

Catalog number: A18944.30
250 g, Each
Thermo Scientific Chemicals

Dimethyl maleate, 96%, Thermo Scientific Chemicals

Catalog number: A18944.30
250 g, Each
Quantity
Catalog number: A18944.30
also known as A18944-30
Price (USD)
Price: 43.60
Online price: 37.65
Your price:
Quantity
-

Chemical Identifiers

CAS
624-48-6
IUPAC Name
1,4-dimethyl (2Z)-but-2-enedioate
Molecular Formula
C6H8O4
InChI Key
LDCRTTXIJACKKU-ARJAWSKDSA-N
SMILES
COC(=O)\C=C/C(=O)OC
Appearance (Color)
Clear, colourless
Assay (GC)
> 95.0%
Refractive Index
1.4390 - 1.4450 @20?C
Form
liquid

Description

Dimethyl maleate is acts as a dienophile involved in ultrasonic irradiation promoted Diels-Alder reaction with substituted furans. It is widely used as an intermediate and additive for pharmaceuticals, copolymers, pigments, plastics, paints, adhesives and agrochemicals. As an internal modifier, it increases the glass transition temperature of styrene or vinyl chloride polymer. It plays a vital role to improve the hardness and toughness of polymer films such as copolymers of vinyl acetate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimethyl maleate is acts as a dienophile involved in ultrasonic irradiation promoted Diels-Alder reaction with substituted furans. It is widely used as an intermediate and additive for pharmaceuticals, copolymers, pigments, plastics, paints, adhesives and agrochemicals. As an internal modifier, it increases the glass transition temperature of styrene or vinyl chloride polymer. It plays a vital role to improve the hardness and toughness of polymer films such as copolymers of vinyl acetate.

Solubility
Miscible with water.

Notes
Incompatible with acids, bases, oxidizing agents and reducing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text