Thermo Scientific Chemicals

Anthrone, 95%, Thermo Scientific Chemicals

Catalog number: A19118.22
100 g, Each
Thermo Scientific Chemicals

Anthrone, 95%, Thermo Scientific Chemicals

Catalog number: A19118.22
100 g, Each
Quantity
Catalog number: A19118.22
also known as A19118-22
Price (USD)
Price: 237.00
Online price: 201.65
Your price:
Quantity
-

Chemical Identifiers

CAS
90-44-8
IUPAC Name
9,10-dihydroanthracen-9-one
Molecular Formula
C14H10O
InChI Key
RJGDLRCDCYRQOQ-UHFFFAOYSA-N
SMILES
O=C1C2=CC=CC=C2CC2=CC=CC=C12
Form
Crystalline powder or needles
Appearance (Color)
Cream to yellow to pale brown
Assay (GC)
≥94.0%
Identification (FTIR)
Conforms

Description

It finds its applications in organic synthesis. It is used for a popular cellulose assay and in the colorometric determination of carbohydrates in body fluids and of animal starch in liver tissue. The anthrones are used in pharmacy as laxative. They stimulate the motion of the colon and are responsible for less water reabsorption. They may only be used for a short amount of time, because long time use may lead to loss of electrolytes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in alcohol, benzene, and hot NaOH solutions. Insoluble in water

Notes
Light Sensitive. Store away from oxidizing agents and light. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text