Thermo Scientific Chemicals

4-Nitroanisole, 97%, Thermo Scientific Chemicals

250g, Each
Thermo Scientific Chemicals

4-Nitroanisole, 97%, Thermo Scientific Chemicals

250g, Each
Quantity
Catalog number: A19152.30
also known as A19152-30
Price (USD)
Price: 36.70
Your Price:
Quantity
-

Chemical Identifiers

CAS
100-17-4
IUPAC Name
1-methoxy-4-nitrobenzene
Molecular Formula
C7H7NO3
InChI Key
BNUHAJGCKIQFGE-UHFFFAOYSA-N
SMILES
COC1=CC=C(C=C1)[N+]([O-])=O
Appearance (Color)
Cream to yellow to brown
Identification (FTIR)
Conforms
Assay (GC)
≥96.0%
Form
Crystals or powder or crystalline powder or fused/lumpy solid
Melting Point (clear melt)
50-55?C

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only