Geranylacetone, (E)+(Z), 97%, (Z)-isomer (nerylacetone) ca 45%
Geranylacetone, (E)+(Z), 97%, (Z)-isomer (nerylacetone) ca 45%
Thermo Scientific Chemicals

Geranylacetone, (E)+(Z), 97%, (Z)-isomer (nerylacetone) ca 45%

CAS: 689-67-8 | C13H22O | 194.318 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
25 g
100 g
產品號碼 A19184.22
亦稱為 A19184-22
價格 (TWD)
-
申請報價
Quantity:
100 g
Request bulk or custom format
化學識別
CAS689-67-8
IUPAC Name(5E)-6,10-dimethylundeca-5,9-dien-2-one
Molecular FormulaC13H22O
InChI KeyHNZUNIKWNYHEJJ-FMIVXFBMSA-N
SMILESCC(C)=CCC\C(C)=C\CCC(C)=O
檢視更多
規格Specification Sheet規格表
Refractive Index1.4645 - 1.4695 @ 20?C
Appearance (Color)Clear. colorless to pale yellow
FormLiquid
Composition(Geranylacetone) content: ≥ 50.0%
Assay (GC)>96.0% (sum of isomers)
It is used as a flavoring agent and adjuvant. It is also employed as a substance, extract, or preparation for diffusing or imparting an agreeable or attractive smell.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a flavoring agent and adjuvant. It is also employed as a substance, extract, or preparation for diffusing or imparting an agreeable or attractive smell.

Solubility
Not miscible or difficult to mix in water. Soluble in alcohol.

Notes
Store away from oxidizing agents and heat. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep it refregirated.
RUO – Research Use Only

General References:

  1. Andrew J. Simkin, et. al. The tomato carotenoid cleavage dioxygenase 1 genes contribute to the formation of the flavor volatiles b-ionone, pseudoionone, and geranylacetone.The Plant Journal.2004,40882-892.
  2. Friedrich Jüttner. The Algal Excretion Product, Geranylacetone: A Potent Inhibitor of Carotene Biosynthesis in Synechococcus.Pharmacology.1979,34(11), 957-960.