Thermo Scientific Chemicals

p-Cymene, 97%, Thermo Scientific Chemicals

Catalog number: A19226.AP
500 mL, Each
Thermo Scientific Chemicals

p-Cymene, 97%, Thermo Scientific Chemicals

Catalog number: A19226.AP
500 mL, Each
Quantity
Catalog number: A19226.AP
also known as A19226-AP
Price (USD)
Price: 30.90
Online price: 26.65
Your price:
Quantity
-

Chemical Identifiers

CAS
99-87-6
IUPAC Name
1-methyl-4-(propan-2-yl)benzene
Molecular Formula
C10H14
InChI Key
HFPZCAJZSCWRBC-UHFFFAOYSA-N
SMILES
CC(C)C1=CC=C(C)C=C1
Appearance (Color)
Clear colorless
Identification (FTIR)
Conforms
Form
Liquid
Assay (GC)
≥96.0%
Refractive Index
1.4875-1.4925 @ 20?C

Description

It is used in heat-exchange media, metal polishes, and thinners for lacquers/varnishes. Also used to make para-cresol, carvacrol, and synthetic resins. It is also used as a component of commercial terpene solvent mixtures.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in heat-exchange media, metal polishes, and thinners for lacquers/varnishes. Also used to make para-cresol, carvacrol, and synthetic resins. It is also used as a component of commercial terpene solvent mixtures.

Solubility
Soluble in water at 0.023g/L.20°C.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text