Thermo Scientific Chemicals

Decafluorobenzophenone, 97%, Thermo Scientific Chemicals

Catalog number: A19262.03
1 g, Each
Thermo Scientific Chemicals

Decafluorobenzophenone, 97%, Thermo Scientific Chemicals

Catalog number: A19262.03
1 g, Each
Quantity
Catalog number: A19262.03
also known as A19262-03
Price (USD)
Price: 54.20
Online price: 46.65
Your price:
Quantity
-

Chemical Identifiers

CAS
853-39-4
IUPAC Name
bis(2,3,4,5,6-pentafluorophenyl)methanone
Molecular Formula
C13F10O
InChI Key
WWQLXRAKBJVNCC-UHFFFAOYSA-N
SMILES
FC1=C(F)C(F)=C(C(=O)C2=C(F)C(F)=C(F)C(F)=C2F)C(F)=C1F
Melting Point (clear melt)
89.0-95.0°C
Form
Crystals or powder or crystalline powder or lumps or fused solid
Appearance (Color)
White
Assay (GC)
≥96.0%

Description

Perfluorobenzophenone was used in the synthesis of fluorinated acridones and acridines. Fluorinated polyetherketones prepared from decafluorobenzophenone. Laser pulse excitation of decafluorobenzophenone (DFB) in acetonitrile yields the triplet 3DFB.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Perfluorobenzophenone was used in the synthesis of fluorinated acridones and acridines. Fluorinated polyetherketones prepared from decafluorobenzophenone. Laser pulse excitation of decafluorobenzophenone (DFB) in acetonitrile yields the triplet 3DFB.

Solubility
Soluble in methanol. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text