cis-4-Hexen-1-ol, 97%
cis-4-Hexen-1-ol, 97%
cis-4-Hexen-1-ol, 97%
Thermo Scientific Chemicals

cis-4-Hexen-1-ol, 97%

CAS: 928-91-6 | C6H12O | 100.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A19266.14
also known as A19266-14
Price (EUR)
137,00
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
137,00
Each
Chemical Identifiers
CAS928-91-6
IUPAC Name(4Z)-hex-4-en-1-ol
Molecular FormulaC6H12O
InChI KeyVTIODUHBZHNXFP-IHWYPQMZSA-N
SMILESC\C=C/CCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
FormLiquid
Refractive Index1.4410-1.4460 @ 20?C
Appearance (Color)Clear colorless
Assay (GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only