Thermo Scientific Chemicals

Farnesol, mixture of isomers, 96%, Thermo Scientific Chemicals

Catalog number: A19316.22
100 g, Each
Thermo Scientific Chemicals

Farnesol, mixture of isomers, 96%, Thermo Scientific Chemicals

Catalog number: A19316.22
100 g, Each
Quantity
Catalog number: A19316.22
also known as A19316-22
Price (USD)
Price: 158.00
Online price: 134.65
Your price:
Quantity
-

Chemical Identifiers

CAS
4602-84-0
Assay (GC)
≥95.0% (sum of isomers)
GC/MS Analysis
Confirms/identifies presence of 3 isomers
Form
Liquid
Comment
Other isomers: 2(E),6(Z)- and 2(Z),6(E)-
Refractive Index
1.4860-1.4920 @ 20?C

Description

It can be used to induce apoptosis in cell cultures. It is also used as an antimicrobial agent. It is used in perfumes and as a flavoring agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It can be used to induce apoptosis in cell cultures. It is also used as an antimicrobial agent. It is used in perfumes and as a flavoring agent.

Solubility
Not miscible or difficult to mix in water.

Notes
Light sensitive. Store away from oxidizing agents and light. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text