Methyl 4-iodobenzoate, 98%
Methyl 4-iodobenzoate, 98%
Methyl 4-iodobenzoate, 98%
Methyl 4-iodobenzoate, 98%
Thermo Scientific Chemicals

Methyl 4-iodobenzoate, 98%

CAS: 619-44-3 | C8H7IO2 | 262.046 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number A19342.09
also known as A19342-09
Price (EUR)
41,70
Each
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (EUR)
41,70
Each
Add to cart
Chemical Identifiers
CAS619-44-3
IUPAC Namemethyl 4-iodobenzoate
Molecular FormulaC8H7IO2
InChI KeyDYUWQWMXZHDZOR-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC=C(I)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to orange to pale brown
Melting Point (clear melt)113-118?C
Assay (GC)≥97.5%
FormCrystals or powder or crystalline powder or chunks
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only