2,4,6-Tribromoresorcinol, 98%
2,4,6-Tribromoresorcinol, 98%
2,4,6-Tribromoresorcinol, 98%
2,4,6-Tribromoresorcinol, 98%
Thermo Scientific Chemicals

2,4,6-Tribromoresorcinol, 98%

CAS: 2437-49-2 | C6H3Br3O2 | 346.8 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number A19436.03
also known as A19436-03
Price (EUR)
15,40
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (EUR)
15,40
Each
Add to cart
Chemical Identifiers
CAS2437-49-2
IUPAC Name2,4,6-tribromobenzene-1,3-diol
Molecular FormulaC6H3Br3O2
InChI KeyYADZSMVDNYXOOB-UHFFFAOYSA-N
SMILESOC1=C(Br)C(O)=C(Br)C=C1Br
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)108.0-114.0?C
FormPowder
Appearance (Color)Pale pink
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only