Thermo Scientific Chemicals

Diethylbenzene, mixture of isomers, Thermo Scientific Chemicals

Catalog number: A19548.AE
100 mL, Each
Thermo Scientific Chemicals

Diethylbenzene, mixture of isomers, Thermo Scientific Chemicals

Catalog number: A19548.AE
100 mL, Each
Quantity
Catalog number: A19548.AE
also known as A19548-AE
Price (USD)
Price: 20.50
Online price: 15.65
Your price:
Quantity
-

Chemical Identifiers

CAS
25340-17-4
IUPAC Name
1,2-diethylbenzene
Molecular Formula
C10H14
InChI Key
KVNYFPKFSJIPBJ-UHFFFAOYSA-N
SMILES
CCC1=CC=CC=C1CC
Refractive Index
1.4925-1.4985 @ 20°C
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Assay (GC)
≥95.0% (sum of isomers)
Identification (FTIR)
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text