Thermo Scientific Chemicals

1-Bromo-4-chloro-2-fluorobenzene, 98%, Thermo Scientific Chemicals

Catalog number: A19570.14
25 g, Each
Thermo Scientific Chemicals

1-Bromo-4-chloro-2-fluorobenzene, 98%, Thermo Scientific Chemicals

Catalog number: A19570.14
25 g, Each
Quantity
Catalog number: A19570.14
also known as A19570-14
Price (USD)
Price: 147.00
Online price: 126.65
Your price:
Quantity
-

Chemical Identifiers

CAS
1996-29-8
IUPAC Name
1-bromo-4-chloro-2-fluorobenzene
Molecular Formula
C6H3BrClF
InChI Key
FPNVMCMDWZNTEU-UHFFFAOYSA-N
SMILES
FC1=CC(Cl)=CC=C1Br
Form
Low melting solid or clear colorless liquid
Appearance (Color)
White/colorless to pale yellow
Refractive Index
1.5520-1.5560 @ 20?C (as melt)
Assay (GC)
≥97.5%
Identification (FTIR)
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text