gamma-Hexanolactone, 98%
gamma-Hexanolactone, 98%
gamma-Hexanolactone, 98%
gamma-Hexanolactone, 98%
Thermo Scientific Chemicals

gamma-Hexanolactone, 98%

CAS: 695-06-7 | C6H10O2 | 114.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number A19642.14
also known as A19642-14
Price (EUR)
41,30
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (EUR)
41,30
Each
Add to cart
Chemical Identifiers
CAS695-06-7
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Refractive Index1.4370-1.4410 @ 20?C
Appearance (Color)Clear colorless
Assay (GC)≥97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only