Farnesyl acetate, mixture of isomers, 96%
Farnesyl acetate, mixture of isomers, 96%
Farnesyl acetate, mixture of isomers, 96%
Thermo Scientific Chemicals

Farnesyl acetate, mixture of isomers, 96%

CAS: 29548-30-9 | C17H28O2 | 264.409 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number A19778.14
also known as A19778-14
Price (EUR)
66,40
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
66,40
Each
Chemical Identifiers
CAS29548-30-9
IUPAC Name(2E,6E)-3,7,11-trimethyldodeca-2,6,10-trien-1-yl acetate
Molecular FormulaC17H28O2
InChI KeyZGIGZINMAOQWLX-NCZFFCEISA-N
SMILESCC(C)=CCC\C(C)=C\CC\C(C)=C\COC(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Refractive Index1.4750-1.4810 @ 20?C
Appearance (Color)Clear colorless
Assay (GC)≥95.0% (sum of isomers)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only