3,4-Dimethyl-1-pentyn-3-ol, 94%
3,4-Dimethyl-1-pentyn-3-ol, 94%
3,4-Dimethyl-1-pentyn-3-ol, 94%
Thermo Scientific Chemicals

3,4-Dimethyl-1-pentyn-3-ol, 94%

CAS: 1482-15-1 | C7H12O | 112.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A19906.14
also known as A19906-14
Price (EUR)
127,00
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
127,00
Each
Chemical Identifiers
CAS1482-15-1
IUPAC Name3,4-dimethylpent-1-yn-3-ol
Molecular FormulaC7H12O
InChI KeyDZNLEQBXXLGELU-UHFFFAOYNA-N
SMILESCC(C)C(C)(O)C#C
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Refractive Index1.4310-1.4380 @ 20?C
Appearance (Color)Clear colorless
Assay (GC)≥92.0%
3,4-Dimethyl-1-pentyn-3-ol is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4-Dimethyl-1-pentyn-3-ol is used as pharmaceutical intermediates.

Solubility
Fully miscible in water.

Notes
Incompatible with oxidizing agents. Store in a cool, dry conditions in a well sealed container. Store at room temperature.
RUO – Research Use Only