Sodium 3-bromopropanesulfonate, 97%
Sodium 3-bromopropanesulfonate, 97%
Sodium 3-bromopropanesulfonate, 97%
Sodium 3-bromopropanesulfonate, 97%
Thermo Scientific Chemicals

Sodium 3-bromopropanesulfonate, 97%

CAS: 55788-44-8 | C3H7BrNaO3S | 226.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A19930.14
also known as A19930-14
Price (EUR)
115,00
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (EUR)
115,00
Each
Add to cart
Chemical Identifiers
CAS55788-44-8
IUPAC Name3-bromopropane-1-sulfonic acid sodium
Molecular FormulaC3H7BrNaO3S
InChI KeyONPNLMMBTJZJRQ-UHFFFAOYSA-N
SMILES[Na].OS(=O)(=O)CCCBr
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
Assay (unspecified)≥96.0%
Identification (FTIR)Conforms
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only