4-Amino-2-(methylthio)pyrimidine-5-carbonitrile, 97%
4-Amino-2-(methylthio)pyrimidine-5-carbonitrile, 97%
4-Amino-2-(methylthio)pyrimidine-5-carbonitrile, 97%
Thermo Scientific Chemicals

4-Amino-2-(methylthio)pyrimidine-5-carbonitrile, 97%

CAS: 770-30-9 | C6H6N4S | 166.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A19949.14
also known as A19949-14
Price (EUR)
241,00
Each
Quantity:
25 g
Request bulk or custom format
Price (EUR)
241,00
Each
Chemical Identifiers
CAS770-30-9
IUPAC Name4-amino-2-(methylsulfanyl)pyrimidine-5-carbonitrile
Molecular FormulaC6H6N4S
InChI KeyVYDHBDPZNLVELI-UHFFFAOYSA-N
SMILESCSC1=NC=C(C#N)C(N)=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)232.0-241.0?C
Appearance (Color)White to cream to pale yellow
Assay (Non-aqueous acid-base Titration)≥96.0 to ≤104.0%
FormPowder
It is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate.

Solubility
Soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only