A2 and A5 Large Gel Electrophoresis System Standard Comb
Actual product may vary
A2 and A5 Large Gel Electrophoresis System Standard Comb
A2 and A5 Large Gel Electrophoresis System Standard Comb
Thermo Scientific™
A2 and A5 Large Gel Electrophoresis System Standard Comb
Eliminate comb assembly with this assortment of heavy-duty, one-piece combs, for use with Fisherbrand™ A2 and A5 Large Gel Electrophoresis Systems.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeTeeth CountThickness (Metric) Teeth
A2-32CStandard Comb321 mm
A2-12CStandard Comb121 mm
A2-12DStandard Comb121.5 mm
A2-16CStandard Comb161 mm
A2-16DStandard Comb161.5 mm
A2-20CStandard Comb201 mm
A2-20DStandard Comb201.5 mm
A2-24CStandard Comb241 mm
A2-24DStandard Comb241.5 mm
A2-28CStandard Comb281 mm
A2-36CStandard Comb361 mm
A2-8CStandard Comb81 mm
A2-8DStandard Comb81.5 mm
A2-MT2CMicrowell Comb42 (2X)1 mm
A2-MT2DMicrowell Comb42 (2X)1.5 mm
A2-MTCMicrowell Comb211 mm
A2-MTDMicrowell Comb211.5 mm
A2-PREPPrep Comb21.5 mm
A2-WALLWall Comb11.5 mm
A2-RL-18DRapid Load Comb181.5 mm
Catalog number A2-32C
Price (MXN)
-
Type:
Standard Comb
Teeth Count:
32
Thickness (Metric) Teeth:
1 mm
Eliminate comb assembly with an assortment of heavy-duty, one-piece Fisherbrand™ A2 and A5 Large Gel Electrophoresis Systems Combs, for use with Owl A2 and A5 Large Gel Electrophoresis Systems.
Heavy-duty, one-piece combs eliminate comb assembly.
Specifications
Teeth Count32
Thickness (Metric) Teeth1 mm
For Use With (Equipment)Fisherbrand™ A2 and A5 Large Gel Electrophoresis Systems
TypeStandard Comb
Unit SizeEach