A2 and A5 Large Gel System Replacement End Gates
Actual product may vary
A2 and A5 Large Gel System Replacement End Gates
A2 and A5 Large Gel System Replacement End Gates
A2 and A5 Large Gel System Replacement End Gates
Thermo Scientific™
A2 and A5 Large Gel System Replacement End Gates
Use these gasketed UVT gel trays, replacement gaskets and end gates with Thermo Scientific™ A2 and A5 Large Gel Systems.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
A2-GKGasket
A2-EGEnd Gates
A2-UVTGel Tray
Catalog number A2-GK
Price (USD)
25.20
Online Exclusive
28.00
Save 2.80 (10%)
Pack of 1
Type:
Gasket
Price (USD)
25.20
Online Exclusive
28.00
Save 2.80 (10%)
Pack of 1
Use Thermo Scientific™ A2 and A5 Large Gel System Replacement Parts with Owl A2 and A5 Large Gel Systems.
  • UVT gel tray incorporates gasketed end gates which provide a leakproof seal without tape
  • Comb slots are positioned to allow 2, 3 or 4 sample sets to run equal lengths on the same gel
Specifications
Quantity1 Pair
For Use With (Equipment)Owl A2 and A5 Large Gel Electrophoresis System
Product LineOwl
TypeGasket
Unit SizePack of 1