Owl™ A2-OK Multiple Gel Electrophoresis System
Thermo Scientific™

Owl™ A2-OK Multiple Gel Electrophoresis System

Run multiple gels simultaneously with Thermo Scientific™ Owl™ A2-OK Multiple Gel Electrophoresis Systems, which are also ideal for teaching labs.
Have Questions?
Catalog NumberGel Size
A2-OK7 x 8 cm
Catalog number A2-OK
Price (USD)
2,709.00
Online exclusive
3,010.00
Save 301.00
Each
Gel Size:
7 x 8 cm
Price (USD)
2,709.00
Online exclusive
3,010.00
Save 301.00
Each
Run multiple gels simultaneously with Thermo Scientific™ Owl™ A2-OK Multiple Gel Electrophoresis Systems, which are also ideal for teaching labs.
Key Features:
  • Up to six 7 x 8cm minigels can be run on one system
  • Screen up to 120 samples at once with supplied combs
  • Thermo Scientific™ EasyCast™ casting design permits casting without tape or plugs
  • UV-transmissible (UVT) gel trays may be placed directly on the transilluminator for easy photodocumentation
  • Teacher can instruct up to 12 students using one system and one power supply

Includes:

Buffer chamber with three-point leveling base;Thermo Scientific™ SuperSafe™ lid with attached power supply leads; Six EasyCast UVT gasketed gel trays; Twelve 10-well, double-sided combs, 1.0/1.5mm thick; Two external multiple gel casters; Gel tray holder

Specifications
Certifications/ComplianceCE marked
DescriptionGel Size: Up to six 7 x 8cm; Buffer volume: 2.3L; L x W x H: 37.5 × 32.5 × 10.5cm
Dimensions (L x W x H)14.8 x 12.8 x 4.1 in. (37.5 x 32.5 x 10.5 cm)
Height (English)4.1 in.
Height (Metric)10.5 cm
Length (English)14.8 in.
Length (Metric)37.5 cm
Length (Metric) Gel8 cm
Sample Size5 to 144
Volume (Metric) Buffer2.3 L
Width (English)12.8 in.
Width (Metric)32.5 cm
Width (Metric) Gel7 cm
Gel Size7 x 8 cm
Product LineOwl
TypeMultiple Gel Electrophoresis System
Unit SizeEach

Frequently asked questions (FAQs)

Are Bio-Rad power supplies compatible with Owl Electrophoresis systems?

The Owl Electrophoresis Systems are supplied with standard power leads, but we have not tested compatibility of the Owl systems with the Bio-Rad power supplies. For best performance, we recommend using one of our Owl or PowerEase power supplies for the Owl Electrophoresis Systems.