Alexa Fluor™ 750 NHS Ester (Succinimidyl Ester)
Invitrogen™

Alexa Fluor™ 750 NHS Ester (Succinimidyl Ester)

Alexa Fluor™ 750 is a bright and photostable near-IR dye that is spectrally similar to Cy7. Used for stable signalRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
A200111 mg
A375753 x 100 μg
A201115 mg
A3756825 mg
Catalog number A20011
Price (USD)
531.00
Each
Quantity:
1 mg
Price (USD)
531.00
Each

Alexa Fluor™ 750 is a bright and photostable near-IR dye that is spectrally similar to Cy7. Used for stable signal generation in imaging and flow cytometry, Alexa Fluor™ 750 dye is water soluble and pH-insensitive from pH 4 to pH 10. Fluorescence of this long-wavelength Alexa Fluor™ dye is not visible to the human eye but is readily detected by most imaging systems. In addition to reactive dye formulations, we offer Alexa Fluor™ 750 dye conjugated to a variety of antibodies, peptides, proteins, tracers, and amplification substrates optimized for cellular labeling and detection (learn more).

The NHS ester (or succinimidyl ester) of Alexa Fluor™ 750 is the most popular tool for conjugating this dye to a protein or antibody. NHS esters can be used to label to the primary amines (R-NH2) of proteins, amine-modified oligonucleotides, and other amine-containing molecules. The resulting Alexa Fluor™ conjugate will exhibit brighter fluorescence and greater photostability than the conjugates of other spectrally similar fluorophores.

Detailed information about this AlexaFluor™ NHS ester:
Fluorophore label: Alexa Fluor™ 750 dye
Reactive group: NHS ester
Reactivity: Primary amines on proteins and ligands, amine-modified oligonucleotides
Ex/Em of the conjugate: 753/782 nm
Extinction coefficient: 290,000 cm-1M-1
Spectrally similar dyes: Cy7
Molecular weight: ∼1300

Typical Conjugation Reaction
You can conjugate amine-reactive reagents with virtually any protein or peptide (the provided protocol is optimized for IgG antibodies). You can scale the reaction for any amount of protein, but the concentration of the protein should be at least 2 mg/mL for optimal results. We recommend trying three different degrees of labeling, using three different molar ratios of the reactive reagent to protein.

The Alexa Fluor™ NHS ester is typically dissolved in high-quality anhydrous dimethylformamide (DMF) or dimethylsulfoxide (DMSO) (D12345), and the reaction is carried out in 0.1–0.2 M sodium bicarbonate buffer, pH 8.3, at room temperature for 1 hour. Because the pKa of the terminal amine is lower than that of the lysine epsilon-amino group, you may achieve more selective labeling of the amine terminus using a buffer closer to neutral pH.

Conjugate Purification
Labeled antibodies are typically separated from free Alexa Fluor™ dye using a gel filtration column, such as Sephadex™ G-25, BioGel™ P-30, or equivalent. For much larger or smaller proteins, select a gel filtration media with an appropriate molecular weight cut-off or purify by dialysis. We offer several purification kits optimized for different quantities of antibody conjugate:
Antibody Conjugate Purification Kit for 0.5-1 mg (A33086)
Antibody Conjugate Purification Kit for 20-50 μg (A33087)
Antibody Conjugate Purification kit for 50-100 μg (A33088)

Learn More About Protein and Antibody Labeling
We offer a wide selection of Molecular Probes™ antibody and protein labeling kits to fit your starting material and your experimental setup. See our Antibody Labeling kits or use our Labeling Chemistry Selection Tool for other choices. To learn more about our labeling kits, read Kits for Labeling Proteins and Nucleic Acids—Section 1.2 in The Molecular Probes™ Handbook.

We’ll Make a Custom Conjugate for You
If you can’t find what you’re looking for in our online catalog, we’ll prepare a custom antibody or protein conjugate for you. Our custom conjugation service is efficient and confidential, and we stand by the quality of our work. We are ISO 13485:2000 certified.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Chemical ReactivityAmine
Emission782 nm
Excitation753 nm
Label or DyeAlexa Fluor™ 750
Product TypeDye
Quantity1 mg
Reactive MoietyActive Ester, Succinimidyl Ester
Shipping ConditionRoom Temperature
Label TypeAlexa Fluor
Product LineAlexa Fluor
Unit SizeEach
Contents & Storage
Store in freezer (-5 to -30°C) and protect from light.

Frequently asked questions (FAQs)

I am labeling a protein with Alexa Fluor 488 SDP ester. The manual recommends using a sodium bicarbonate buffer at pH 8.3. Can I use a different buffer instead?

Yes. The important thing is to use a buffered solution with a pH between 8.0 and 8.5. Do not use Tris buffer, which has amine groups. Most other buffers will work fine in that pH range. This is also true for other amine-reactive dyes, such as succinimidyl (NHS) esters or TFP esters.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

I am not going to use all of my Alexa Fluor succinimidyl ester reactive dye. Can I just make it up in DMSO and store aliquots at -20 degrees C?

This is not recommended. Any trace amounts of water in the DMSO can promote spontaneous hydrolysis over time. Even if using anhydrous DMSO, DMSO is hygroscopic; it readily absorbs moisture from the atmosphere over time. A better alternative is to dissolve the reactive dye in a volatile solvent, make smaller aliquots and then evaporate off the solvent using a vacuum pump. The smaller aliquots of solid reactive dye should then be stored frozen, desiccated and protected from light. Contact Technical Support by sending an email to techsupport@thermofisher.com for the recommended volatile solvent.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Citations & References (22)

Citations & References
Abstract
Tyramide signal amplification for analysis of kinase activity by intracellular flow cytometry.
Authors:Clutter MR, Heffner GC, Krutzik PO, Sachen KL, Nolan GP,
Journal:Cytometry A
PubMed ID:20824632
'Intracellular flow cytometry permits quantitation of diverse molecular targets at the single-cell level. However, limitations in detection sensitivity inherently restrict the method, sometimes resulting in the inability to measure proteins of very low abundance or to differentiate cells expressing subtly different protein concentrations. To improve these measurements, an enzymatic amplification ... More
Quantitative comparison of long-wavelength Alexa Fluor dyes to Cy dyes: fluorescence of the dyes and their bioconjugates.
Authors:Berlier JE, Rothe A, Buller G, Bradford J, Gray DR, Filanoski BJ, Telford WG, Yue S, Liu J, Cheung CY, Chang W, Hirsch JD, Beechem JM, Haugland RP, Haugland RP
Journal:J Histochem Cytochem
PubMed ID:14623938
'Amine-reactive N-hydroxysuccinimidyl esters of Alexa Fluor fluorescent dyes with principal absorption maxima at about 555 nm, 633 nm, 647 nm, 660 nm, 680 nm, 700 nm, and 750 nm were conjugated to antibodies and other selected proteins. These conjugates were compared with spectrally similar protein conjugates of the Cy3, Cy5, ... More
Fluorescent cell barcoding in flow cytometry allows high-throughput drug screening and signaling profiling.
Authors:Krutzik PO, Nolan GP
Journal:Nat Methods
PubMed ID:16628206
'Flow cytometry allows high-content, multiparameter analysis of single cells, making it a promising tool for drug discovery and profiling of intracellular signaling. To add high-throughput capacity to flow cytometry, we developed a cell-based multiplexing technique called fluorescent cell barcoding (FCB). In FCB, each sample is labeled with a different signature, ... More
Noninvasive tracking of donor cell homing by near-infrared fluorescence imaging shortly after bone marrow transplantation.
Authors:Ushiki T, Kizaka-Kondoh S, Ashihara E, Tanaka S, Masuko M, Hirai H, Kimura S, Aizawa Y, Maekawa T, Hiraoka M,
Journal:PLoS One
PubMed ID:20559437
'BACKGROUND: Many diseases associated with bone marrow transplantation (BMT) are caused by transplanted hematopoietic cells, and the onset of these diseases occurs after homing of donor cells in the initial phase after BMT. Noninvasive observation of donor cell homing shortly after transplantation is potentially valuable for improving therapeutic outcomes of ... More
Evaluation of fluorophores for optimal performance in localization-based super-resolution imaging.
Authors:Dempsey GT, Vaughan JC, Chen KH, Bates M, Zhuang X,
Journal:Nat Methods
PubMed ID:22056676
'One approach to super-resolution fluorescence imaging uses sequential activation and localization of individual fluorophores to achieve high spatial resolution. Essential to this technique is the choice of fluorescent probes; the properties of the probes, including photons per switching event, on-off duty cycle, photostability and number of switching cycles, largely dictate ... More