Alexa Fluor™ 488 NHS Ester (Succinimidyl Ester)
Alexa Fluor™ 488 NHS Ester (Succinimidyl Ester)
Invitrogen™

Alexa Fluor™ 488 NHS Ester (Succinimidyl Ester)

Alexa Fluor™ 488 is a bright, green-fluorescent dye with excitation ideally suited for the 488 nm laser line. Used forRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
A201005 mg
A200001 mg
Catalog number A20100
Price (USD)
918.00
Each
Add to cart
Quantity:
5 mg
Price (USD)
918.00
Each
Add to cart

Alexa Fluor™ 488 is a bright, green-fluorescent dye with excitation ideally suited for the 488 nm laser line. Used for stable signal generation in imaging and flow cytometry, Alexa Fluor™ 488 dye is water soluble and pH-insensitive from pH 4 to pH 10. In addition to reactive dye formulations, we offer Alexa Fluor™ 488 dye conjugated to a variety of antibodies, peptides, proteins, tracers, and amplification substrates optimized for cellular labeling and detection (learn more).

The NHS ester (or succinimidyl ester) of Alexa Fluor™ 488 is the most popular tool for conjugating this dye to a protein or antibody. NHS esters can be used to label to the primary amines (R-NH2) of proteins, amine-modified oligonucleotides, and other amine-containing molecules. The resulting Alexa Fluor™ conjugate will exhibit brighter fluorescence and greater photostability than the conjugates of other spectrally similar fluorophores.

Detailed information about this AlexaFluor™ NHS ester:
Fluorophore label: Alexa Fluor™ 488 dye
Reactive group: NHS ester
Reactivity: Primary amines on proteins and ligands, amine-modified oligonucleotides
Ex/Em of the conjugate: 494/517 nm
Extinction coefficient: 73,000 cm-1M-1
Spectrally similar dyes: FITC, Oregon Green 488
Molecular weight: 643.4

Typical Conjugation Reaction
You can conjugate amine-reactive reagents with virtually any protein or peptide (the provided protocol is optimized for IgG antibodies). You can scale the reaction for any amount of protein, but the concentration of the protein should be at least 2 mg/mL for optimal results. We recommend trying three different degrees of labeling, using three different molar ratios of the reactive reagent to protein.

The Alexa Fluor™ NHS ester is typically dissolved in high-quality anhydrous dimethylformamide (DMF) or dimethylsulfoxide (DMSO) (D12345), and the reaction is carried out in 0.1–0.2 M sodium bicarbonate buffer, pH 8.3, at room temperature for 1 hour. Because the pKa of the terminal amine is lower than that of the lysine epsilon-amino group, you may achieve more selective labeling of the amine terminus using a buffer closer to neutral pH.

Conjugate Purification
Labeled antibodies are typically separated from free Alexa Fluor™ dye using a gel filtration column, such as Sephadex™ G-25, BioGel™ P-30, or equivalent. For much larger or smaller proteins, select a gel filtration media with an appropriate molecular weight cut-off or purify by dialysis. We offer several purification kits optimized for different quantities of antibody conjugate:
Antibody Conjugate Purification Kit for 0.5-1 mg (A33086)
Antibody Conjugate Purification Kit for 20-50 μg (A33087)
Antibody Conjugate Purification kit for 50-100 μg (A33088)

Learn More About Protein and Antibody Labeling
We offer a wide selection of Molecular Probes™ antibody and protein labeling kits to fit your starting material and your experimental setup. See our Antibody Labeling kits or use our Labeling Chemistry Selection Tool for other choices. To learn more about our labeling kits, read Kits for Labeling Proteins and Nucleic Acids—Section 1.2 in The Molecular Probes™ Handbook.

We’ll Make a Custom Conjugate for You
If you can’t find what you’re looking for in our online catalog, we’ll prepare a custom antibody or protein conjugate for you. Our custom conjugation service is efficient and confidential, and we stand by the quality of our work. We are ISO 13485:2000 certified.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Chemical ReactivityAmine
Emission517 nm
Excitation494 nm
Label or DyeAlexa Fluor™ 488
Product TypeDye
Quantity5 mg
Reactive MoietyActive Ester, TFP Ester
Shipping ConditionRoom Temperature
Label TypeAlexa Fluor
Product LineAlexa Fluor
Unit SizeEach
Contents & Storage
Store in freezer (-5 to -30°C) and protect from light.

Frequently asked questions (FAQs)

I am labeling a protein with Alexa Fluor 488 SDP ester. The manual recommends using a sodium bicarbonate buffer at pH 8.3. Can I use a different buffer instead?

Yes. The important thing is to use a buffered solution with a pH between 8.0 and 8.5. Do not use Tris buffer, which has amine groups. Most other buffers will work fine in that pH range. This is also true for other amine-reactive dyes, such as succinimidyl (NHS) esters or TFP esters.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

I am not going to use all of my Alexa Fluor succinimidyl ester reactive dye. Can I just make it up in DMSO and store aliquots at -20 degrees C?

This is not recommended. Any trace amounts of water in the DMSO can promote spontaneous hydrolysis over time. Even if using anhydrous DMSO, DMSO is hygroscopic; it readily absorbs moisture from the atmosphere over time. A better alternative is to dissolve the reactive dye in a volatile solvent, make smaller aliquots and then evaporate off the solvent using a vacuum pump. The smaller aliquots of solid reactive dye should then be stored frozen, desiccated and protected from light. Contact Technical Support by sending an email to techsupport@thermofisher.com for the recommended volatile solvent.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

What are the signal intensity differences between Alexa Fluor 350 dye and Alexa Fluor 488 dye?

In general, blue fluorescent dyes are not as bright as other dyes further along the color spectrum. Blue dyes are structurally smaller and have lower extinction coefficients, so they are typically not as bright compared to the green, red, and far red dyes.
When using an Alexa Fluor 350 secondary antibody, we recommend that you use it for highly expressed targets and at a higher concentration than what is typically required for green or red secondary antibodies.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Citations & References (198)

Citations & References
Abstract
Activity of human IgG and IgA subclasses in immune defense against Neisseria meningitidis serogroup B.
Authors:Vidarsson G,van Der Pol WL,van Den Elsen JM,Vilé H,Jansen M,Duijs J,Morton HC,Boel E,Daha MR,Corthésy B,van De Winkel JG
Journal:Journal of immunology (Baltimore, Md. : 1950)
PubMed ID:11342648
Reconstituted syntaxin1a/SNAP25 interacts with negatively charged lipids as measured by lateral diffusion in planar supported bilayers.
Authors:Wagner ML, Tamm LK
Journal:Biophys J
PubMed ID:11423412
According to the soluble N-ethylmaleimide-sensitive factor (NSF)-attachment protein (SNAP) receptor hypothesis (SNARE hypothesis), interactions between target SNAREs and vesicle SNAREs (t- and v-SNAREs) are required for membrane fusion in intracellular vesicle transport and exocytosis. The precise role of the SNAREs in tethering, docking, and fusion is still disputed. Biophysical measurements ... More
Tubulin equilibrium unfolding followed by time-resolved fluorescence and fluorescence correlation spectroscopy.
Authors:Sánchez SA, Brunet JE, Jameson DM, Lagos R, Monasterio O
Journal:Protein Sci
PubMed ID:14691224
The pathway for the in vitro equilibrium unfolding of the tubulin heterodimer by guanidinium chloride (GdmCl) has been studied using several spectroscopic techniques, specifically circular dichroism (CD), two-photon Fluorescence Correlation Spectroscopy (FCS), and time-resolved fluorescence, including lifetime and dynamic polarization. The results show that tubulin unfolding is characterized by distinct ... More
Segregation of nitrogen fixation and oxygenic photosynthesis in the marine cyanobacterium Trichodesmium.
Authors:Berman-Frank I, Lundgren P, Chen YB, Küpper H, Kolber Z, Bergman B, Falkowski P
Journal:Science
PubMed ID:11711677
'In the modern ocean, a significant amount of nitrogen fixation is attributed to filamentous, nonheterocystous cyanobacteria of the genus Trichodesmium. In these organisms, nitrogen fixation is confined to the photoperiod and occurs simultaneously with oxygenic photosynthesis. Nitrogenase, the enzyme responsible for biological N2 fixation, is irreversibly inhibited by oxygen in ... More
Cytoplasmic dynein/dynactin drives kinetochore protein transport to the spindle poles and has a role in mitotic spindle checkpoint inactivation.
Authors:Howell BJ, McEwen BF, Canman JC, Hoffman DB, Farrar EM, Rieder CL, Salmon ED
Journal:J Cell Biol
PubMed ID:11756470
'We discovered that many proteins located in the kinetochore outer domain, but not the inner core, are depleted from kinetochores and accumulate at spindle poles when ATP production is suppressed in PtK1 cells, and that microtubule depolymerization inhibits this process. These proteins include the microtubule motors CENP-E and cytoplasmic dynein, ... More