Alexa Fluor™ 532 NHS Ester (Succinimidyl Ester)
Invitrogen™

Alexa Fluor™ 532 NHS Ester (Succinimidyl Ester)

Alexa Fluor™ 532 is a bright yellow dye with excitation ideally suited for the frequency-doubled Nd:YAG laser line. Used for자세히 알아보기
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호수량
A20101MP
A-20101MP으로도 사용됨
5 mg
A20001
A-20001으로도 사용됨
1 mg
카탈로그 번호 A20101MP
A-20101MP으로도 사용됨
제품 가격(KRW)
2,421,000
온라인 행사
Ends: 30-Jun-2026
2,848,000
할인액 427,000 (15%)
Each
수량:
5 mg
제품 가격(KRW)
2,421,000
온라인 행사
Ends: 30-Jun-2026
2,848,000
할인액 427,000 (15%)
Each

Alexa Fluor™ 532 is a bright yellow dye with excitation ideally suited for the frequency-doubled Nd:YAG laser line. Used for stable signal generation in imaging and flow cytometry, Alexa Fluor™ 532 dye is water soluble and pH-insensitive from pH 4 to pH 10. In addition to reactive dye formulations, we offer Alexa Fluor™ 532 dye conjugated to a variety of antibodies, peptides, proteins, tracers, and amplification substrates optimized for cellular labeling and detection (learn more).

The NHS ester (or succinimidyl ester) of Alexa Fluor™ 532 is the most popular tool for conjugating this dye to a protein or antibody. NHS esters can be used to label to the primary amines (R-NH2) of proteins, amine-modified oligonucleotides, and other amine-containing molecules. The resulting Alexa Fluor™ conjugate will exhibit brighter fluorescence and greater photostability than the conjugates of other spectrally similar fluorophores.

Detailed information about this AlexaFluor™ NHS ester:

  • Fluorophore label: Alexa Fluor™ 532 dye
  • Reactive group: NHS ester
  • Reactivity: Primary amines on proteins and ligands, amine-modified oligonucleotides
  • Ex/Em of the conjugate: 530/555 nm
  • Extinction coefficient: 81,000 cm-1M-1
  • Molecular weight: 723.8

Typical Conjugation Reaction

You can conjugate amine-reactive reagents with virtually any protein or peptide (the provided protocol is optimized for IgG antibodies). You can scale the reaction for any amount of protein, but the concentration of the protein should be at least 2 mg/mL for optimal results. We recommend trying three different degrees of labeling, using three different molar ratios of the reactive reagent to protein.

The Alexa Fluor™ NHS ester is typically dissolved in high-quality anhydrous dimethylformamide (DMF) or dimethylsulfoxide (DMSO) (D12345), and the reaction is carried out in 0.1–0.2 M sodium bicarbonate buffer, pH 8.3, at room temperature for 1 hour. Because the pKa of the terminal amine is lower than that of the lysine epsilon-amino group, you may achieve more selective labeling of the amine terminus using a buffer closer to neutral pH.

Conjugate Purification

Labeled antibodies are typically separated from free Alexa Fluor™ dye using a gel filtration column, such as Sephadex™ G-25, BioGel™ P-30, or equivalent. For much larger or smaller proteins, select a gel filtration media with an appropriate molecular weight cut-off or purify by dialysis. We offer several purification kits optimized for different quantities of antibody conjugate:

  • Antibody Conjugate Purification Kit for 0.5-1 mg (A33086)
  • Antibody Conjugate Purification Kit for 20-50 μg (A33087)
  • Antibody Conjugate Purification kit for 50-100 μg (A33088)

Learn More About Protein and Antibody Labeling

We offer a wide selection of Molecular Probes™ antibody and protein labeling kits to fit your starting material and your experimental setup. See our Antibody Labeling kits or use our Labeling Chemistry Selection Tool for other choices. To learn more about our labeling kits, read Kits for Labeling Proteins and Nucleic Acids—Section 1.2 in The Molecular Probes™ Handbook.

We'll Make a Custom Conjugate for You

If you can't find what you're looking for in our online catalog, we'll prepare a custom antibody or protein conjugate for you. Our custom conjugation service is efficient and confidential, and we stand by the quality of our work. We are ISO 13485:2000 certified.

For Research Use Only. Not for use in diagnostic procedures.
사양
화학물질 반응성Amine
방출555 nm
여기530 nm
라벨 또는 염료Alexa Fluor™ 532
제품 유형Alexa Fluor™ 532 NHS Ester (Succinimidyl Ester)
수량5 mg
반응성 부분Active Ester, Succinimidyl Ester
배송 조건Room Temperature
라벨 유형Alexa Fluor
제품라인Alexa Fluor
Unit SizeEach
구성 및 보관
Store in freezer (-5°C to -30°C) and protect from light.

자주 묻는 질문(FAQ)

I am labeling a protein with Alexa Fluor 488 SDP ester. The manual recommends using a sodium bicarbonate buffer at pH 8.3. Can I use a different buffer instead?

Yes. The important thing is to use a buffered solution with a pH between 8.0 and 8.5. Do not use Tris buffer, which has amine groups. Most other buffers will work fine in that pH range. This is also true for other amine-reactive dyes, such as succinimidyl (NHS) esters or TFP esters.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

I am not going to use all of my Alexa Fluor succinimidyl ester reactive dye. Can I just make it up in DMSO and store aliquots at -20 degrees C?

This is not recommended. Any trace amounts of water in the DMSO can promote spontaneous hydrolysis over time. Even if using anhydrous DMSO, DMSO is hygroscopic; it readily absorbs moisture from the atmosphere over time. A better alternative is to dissolve the reactive dye in a volatile solvent, make smaller aliquots and then evaporate off the solvent using a vacuum pump. The smaller aliquots of solid reactive dye should then be stored frozen, desiccated and protected from light. Contact Technical Support by sending an email to techsupport@thermofisher.com for the recommended volatile solvent.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

인용 및 참조 문헌 (17)

인용 및 참조 문헌
Abstract
Benzodiazepine binding studies on living cells: application of small ligands for fluorescence correlation spectroscopy.
Authors:Hegener O, Jordan R, Häberlein H
Journal:Biol Chem
PubMed ID:12530545
'We demonstrate the applicability of fluorescence correlation spectroscopy (FCS) for receptor binding studies using low molecular weight ligands on the membranes of living nerve cells. The binding of the benzodiazepine Ro 7-1986/602 (N-des-diethyl-fluorazepam), labeled with the fluorophore Alexa 532, to the benzodiazepine receptor was analyzed quantitatively at the membrane of ... More
Lateral mobility and specific binding to GABA(A) receptors on hippocampal neurons monitored by fluorescence correlation spectroscopy.
Authors:Meissner O, Häberlein H
Journal:Biochemistry
PubMed ID:12578381
'The binding behavior of a fluorescently labeled muscimol derivative to the GABA(A) receptor was analyzed at rat hippocampal neurons by fluorescence correlation spectroscopy. After muscimol had been labeled with the fluorophore Alexa Fluor 532, specific binding constants for binding of the dye-labeled ligand (Mu-Alexa) to the GABA(A) receptor were determined. ... More
Dynamics of beta2-adrenergic receptor-ligand complexes on living cells.
Authors:Hegener O, Prenner L, Runkel F, Baader SL, Kappler J, Häberlein H
Journal:Biochemistry
PubMed ID:15147203
'The agonist-induced dynamic regulation of the beta(2)-adrenergic receptor (beta(2)-AR) on living cells was examined by means of fluorescence correlation spectroscopy (FCS) using a fluorescence-labeled arterenol derivative (Alexa-NA) in hippocampal neurons and in alveolar epithelial type II cell line A549. Alexa-NA specifically bound to the beta(2)-AR of neurons with a K(D) ... More
Quantitative screening of single copies of human papilloma viral DNA without amplification.
Authors:Li J, Lee JY, Yeung ES
Journal:Anal Chem
PubMed ID:16970325
'We describe a novel quantitative viral screening method based on single-molecule detection that does not require amplification. DNA of human papilloma virus (HPV), the major etiological agent of cervical cancer, served as the screening target in this study. Eight 100-nucleotide single-stranded DNA probes were designed complementary to the E6-E7 gene ... More
Plasma membrane cholesterol content affects nitric oxide diffusion dynamics and signaling.
Authors:Miersch S, Espey MG, Chaube R, Akarca A, Tweten R, Ananvoranich S, Mutus B,
Journal:J Biol Chem
PubMed ID:18445594
'Nitric oxide (NO) signaling is inextricably linked to both its physical and chemical properties. Due to its preferentially hydrophobic solubility, NO molecules tend to partition from the aqueous milieu into biological membranes. We hypothesized that plasma membrane ordering provided by cholesterol further couples the physics of NO diffusion with cellular ... More