Alexa Fluor™ 568 NHS Ester (Succinimidyl Ester)
Invitrogen™

Alexa Fluor™ 568 NHS Ester (Succinimidyl Ester)

Alexa Fluor™ 568 is a bright orange dye. Used for stable signal generation in imaging and flow cytometry, Alexa Fluor™자세히 알아보기
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호수량
A20103
A-20103으로도 사용됨
5 mg
A200031 mg
카탈로그 번호 A20103
A-20103으로도 사용됨
제품 가격(KRW)
2,360,000
온라인 행사
Ends: 15-Jul-2026
2,776,000
할인액 416,000 (15%)
Each
수량:
5 mg
제품 가격(KRW)
2,360,000
온라인 행사
Ends: 15-Jul-2026
2,776,000
할인액 416,000 (15%)
Each

Alexa Fluor™ 568 is a bright orange dye. Used for stable signal generation in imaging and flow cytometry, Alexa Fluor™ 568 dye is water soluble and pH-insensitive from pH 4 to pH 10. In addition to reactive dye formulations, we offer Alexa Fluor™ 568 dye conjugated to a variety of antibodies, peptides, proteins, tracers, and amplification substrates optimized for cellular labeling and detection (learn more).

The NHS ester (or succinimidyl ester) of Alexa Fluor™ 568 is the most popular tool for conjugating this dye to a protein or antibody. NHS esters can be used to label to the primary amines (R-NH2) of proteins, amine-modified oligonucleotides, and other amine-containing molecules. The resulting Alexa Fluor™ conjugate will exhibit brighter fluorescence and greater photostability than the conjugates of other spectrally similar fluorophores.

Detailed information about this AlexaFluor™ NHS ester:
Fluorophore label: Alexa Fluor™ 568 dye
Reactive group: NHS ester
Reactivity: Primary amines on proteins and ligands, amine-modified oligonucleotides
Ex/Em of the conjugate: 578/602 nm
Extinction coefficient: 88,000 cm-1M-1
Spectrally similar dyes: Rhodamine Red
Molecular weight: 791.8

Typical Conjugation Reaction
You can conjugate amine-reactive reagents with virtually any protein or peptide (the provided protocol is optimized for IgG antibodies). You can scale the reaction for any amount of protein, but the concentration of the protein should be at least 2 mg/mL for optimal results. We recommend trying three different degrees of labeling, using three different molar ratios of the reactive reagent to protein.

The Alexa Fluor™ NHS ester is typically dissolved in high-quality anhydrous dimethylformamide (DMF) or dimethylsulfoxide (DMSO) (D12345), and the reaction is carried out in 0.1–0.2 M sodium bicarbonate buffer, pH 8.3, at room temperature for 1 hour. Because the pKa of the terminal amine is lower than that of the lysine epsilon-amino group, you may achieve more selective labeling of the amine terminus using a buffer closer to neutral pH.

Conjugate Purification
Labeled antibodies are typically separated from free Alexa Fluor™ dye using a gel filtration column, such as Sephadex™ G-25, BioGel™ P-30, or equivalent. For much larger or smaller proteins, select a gel filtration media with an appropriate molecular weight cut-off or purify by dialysis. We offer several purification kits optimized for different quantities of antibody conjugate:
Antibody Conjugate Purification Kit for 0.5-1 mg (A33086)
Antibody Conjugate Purification Kit for 20-50 μg (A33087)
Antibody Conjugate Purification kit for 50-100 μg (A33088)

Learn More About Protein and Antibody Labeling
We offer a wide selection of Molecular Probes™ antibody and protein labeling kits to fit your starting material and your experimental setup. See our Antibody Labeling kits or use our Labeling Chemistry Selection Tool for other choices. To learn more about our labeling kits, read Kits for Labeling Proteins and Nucleic Acids—Section 1.2 in The Molecular Probes™ Handbook.

We’ll Make a Custom Conjugate for You
If you can’t find what you’re looking for in our online catalog, we’ll prepare a custom antibody or protein conjugate for you. Our custom conjugation service is efficient and confidential, and we stand by the quality of our work. We are ISO 13485:2000 certified.

For Research Use Only. Not for use in diagnostic procedures.
사양
화학물질 반응성Amine
방출602 nm
여기578 nm
라벨 또는 염료Alexa Fluor™ 568
제품 유형Dye
수량5 mg
반응성 부분Active Ester, Succinimidyl Ester
배송 조건Room Temperature
라벨 유형Alexa Fluor
제품라인Alexa Fluor
Unit SizeEach
구성 및 보관
Store in freezer (-5 to -30°C) and protect from light.

자주 묻는 질문(FAQ)

I am labeling a protein with Alexa Fluor 488 SDP ester. The manual recommends using a sodium bicarbonate buffer at pH 8.3. Can I use a different buffer instead?

Yes. The important thing is to use a buffered solution with a pH between 8.0 and 8.5. Do not use Tris buffer, which has amine groups. Most other buffers will work fine in that pH range. This is also true for other amine-reactive dyes, such as succinimidyl (NHS) esters or TFP esters.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

I am not going to use all of my Alexa Fluor succinimidyl ester reactive dye. Can I just make it up in DMSO and store aliquots at -20 degrees C?

This is not recommended. Any trace amounts of water in the DMSO can promote spontaneous hydrolysis over time. Even if using anhydrous DMSO, DMSO is hygroscopic; it readily absorbs moisture from the atmosphere over time. A better alternative is to dissolve the reactive dye in a volatile solvent, make smaller aliquots and then evaporate off the solvent using a vacuum pump. The smaller aliquots of solid reactive dye should then be stored frozen, desiccated and protected from light. Contact Technical Support by sending an email to techsupport@thermofisher.com for the recommended volatile solvent.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

인용 및 참조 문헌 (28)

인용 및 참조 문헌
Abstract
Segregation of nitrogen fixation and oxygenic photosynthesis in the marine cyanobacterium Trichodesmium.
Authors:Berman-Frank I, Lundgren P, Chen YB, Küpper H, Kolber Z, Bergman B, Falkowski P
Journal:Science
PubMed ID:11711677
'In the modern ocean, a significant amount of nitrogen fixation is attributed to filamentous, nonheterocystous cyanobacteria of the genus Trichodesmium. In these organisms, nitrogen fixation is confined to the photoperiod and occurs simultaneously with oxygenic photosynthesis. Nitrogenase, the enzyme responsible for biological N2 fixation, is irreversibly inhibited by oxygen in ... More
High density lipoprotein (HDL) particle uptake mediated by scavenger receptor class B type 1 results in selective sorting of HDL cholesterol from protein and polarized cholesterol secretion.
Authors:Silver DL, Wang N, Xiao X, Tall AR
Journal:J Biol Chem
PubMed ID:11301333
'High density lipoprotein (HDL) mediates reverse transport of cholesterol from atheroma foam cells to the liver, but the mechanisms of hepatic uptake and trafficking of HDL particles are poorly understood. In contrast to its accepted role as a cell surface receptor, scavenger receptor class B type 1 (SR-BI) is shown ... More
Acid-induced conformational changes in phosphoglucose isomerase result in its increased cell surface association and deposition on fibronectin fibrils.
Authors:Amraei M, Jia Z, Reboul P, Nabi IR
Journal:J Biol Chem
PubMed ID:12888574
'Phosphoglucose isomerase (PGI) is a glycolytic enzyme that exhibits extracellular cytokine activity as autocrine motility factor, neuroleukin, and maturation factor and that has been recently implicated as an autoantigen in rheumatoid arthritis. In contrast to its receptor-mediated endocytosis at neutral pH, addition of 25 microg/ml of either Alexa 568- or ... More
Serum amyloid A is a ligand for scavenger receptor class B type I and inhibits high density lipoprotein binding and selective lipid uptake.
Authors:Cai L, de Beer MC, de Beer FC, van der Westhuyzen DR
Journal:J Biol Chem
PubMed ID:15561721
'Serum amyloid A is an acute phase protein that is carried in the plasma largely as an apolipoprotein of high density lipoprotein (HDL). In this study we investigated whether SAA is a ligand for the HDL receptor, scavenger receptor class B type I (SR-BI), and how SAA may influence SR-BI-mediated ... More
Differences in endocytosis and intracellular sorting of ricin and viscumin in 3T3 cells.
Authors:Moisenovic M, Tonevitsky A, Agapov I, Niwa H, Schewe H, Bereiter-Hahn J
Journal:Eur J Cell Biol
PubMed ID:12437187
'Ricin and viscumin are heterodimeric protein toxins. Their A-chain is enzymatically active and removes an adenine residue from the 28S rRNA, the B-chain has lectin activity and binds to terminal galactose residues of cell surface receptors. The toxins reveal a high degree of identity in their amino acid sequences. Nevertheless, ... More