Acryloyl-X, SE (6-((acryloyl)amino)hexanoic acid, succinimidyl ester)
Invitrogen™

Acryloyl-X, SE (6-((acryloyl)amino)hexanoic acid, succinimidyl ester)

Acryloyl-X, SE (6-((acryloyl)amino)hexanoic acid, succinimidyl ester) is an amine-reactive molecule that enables crosslinking of acrylamide to proteins and other biomolecules containing amines.
Have Questions?
Catalog NumberQuantity
A207705 mg
Catalog number A20770
Price (USD)
233.00
Each
Quantity:
5 mg
Price (USD)
233.00
Each

Acryloyl-X, SE (6-((acryloyl)amino)hexanoic acid, succinimidyl ester) is an amine-reactive molecule that enables crosslinking of acrylamide to proteins and other biomolecules containing amines. The succinimidyl ester of acryloyl-X covalently reacts with amines of proteins, amine-modified nucleic acids, and other biomolecules to yield acrylamides that can be copolymerized into polyacrylamide matrices or on surfaces, such as in microarrays and in biosensors. For instance, acryloyl-X, SE is used to prepare streptavidin acrylamide (Cat. No. S21379), which can copolymerize with acrylamide on a polymeric surface to create a uniform monolayer of immobilized streptavidin for binding to biotinylated ligands such as hybridization probes, enzymes, antibodies, and drugs.

Acryloyl-X, SE is a critical crosslinking reagent used in the super-resolution imaging techniques expansion microscopy (ExM) and protein-retention expansion microscopy (proExM) to add acrylamides to the amines of proteins of a preserved cell or tissue sample. This crosslinking with acrylamide allows the proteins to be anchored to a polymer hydrogel and polymerized. The polymer hydrogel is then swelled, allowing the cell or tissue sample to be physically expanded to improve spatial resolution in imaging. Expansion microscopy is a versatile super-resolution imaging technique that can be performed on many specimens, including 3D imaging of thick samples. This imaging technique also uses conventional microscopes, enabling easier and faster visualization of nanoscale biological structures without advanced super-resolution microscopes and equipment.

Properties of acryloyl-X, SE:

  • Synonyms – Acryloyl-X, SE; 6-((acryloyl)amino)hexanoic Acid, Succinimidyl Ester; 2,5-Dioxopyrrolidin-1-yl 6-Acrylamidohexanoate; N-Succinimidyl 6-Acrylamidohexanoate; N-[6-[(2,5-Dioxo-1-pyrrolidinyl)oxy]-6-oxohexyl]-2-propenamide
  • Molecular weight (MW) – 282.30
  • Molecular formula – C13H18N2O5
  • CAS number – 63392-86-9

We recommend dissolving in high-quality anhydrous DMSO to attain a stock concentration of up to 10 mM.

For Research Use Only. Not for use in diagnostic procedures.

Specifications
Chemical ReactivityAmine
Label or DyeAcryloyl-X
Product TypeAcryloyl-X SE
Quantity5 mg
Reactive MoietyActive Ester, Succinimidyl Ester
Shipping ConditionRoom Temperature
Label TypeClassic Dyes
Unit SizeEach
Contents & Storage
Store in freezer -5°C to -30°C and protect from light.

Citations & References (7)

Citations & References
Abstract
GelMap: intrinsic calibration and deformation mapping for expansion microscopy.
Authors:Kapitein LC
Journal:Nature methods
PubMed ID:37723243
A highly homogeneous polymer composed of tetrahedron-like monomers for high-isotropy expansion microscopy.
Authors:Boyden ES
Journal:Nature nanotechnology
PubMed ID:33782587
Simultaneous expansion microscopy imaging of proteins and mRNAs via dual-ExM.
Authors:Chang JB
Journal:Scientific reports
PubMed ID:35233025
Protein-retention expansion microscopy of cells and tissues labeled using standard fluorescent proteins and antibodies.
Authors:Boyden ES
Journal:Nature biotechnology
PubMed ID:27376584
Expansion microscopy: principles and uses in biological research.
Authors:Boyden ES
Journal:Nature methods
PubMed ID:30573813