Product Image
Thermo Scientific™
Thermo Scientific™ QSP™ SAV-IT Cap
Thermo Scientific™ QSP™ SAV-IT closures can be pierced for direct tube sampling. Fit virtually all standard tubes
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호마개 크기제품 사이즈
A21079-Q16 mmCase of 10000
A21071-Q12/13 mmCase of 10000
A21071-QPK
A21071-Q으로도 사용됨
12/13 mmPack of 1000
A21079-QPK
A21079-Q으로도 사용됨
16 mmPack of 1000
카탈로그 번호 A21079-Q
제품 가격(KRW)
-
마개 크기:
16 mm
Unit Size:
Case of 10000
Thermo Scientific™ QSP™ SAV-IT closures are made of a proprietary plastic blend and can be pierced for direct tube sampling. Fit virtually all standard tubes, including evacuated blood drawing tubes, glass and plastic test tubes and are great with instrument tubes and sample cups.
Thermo Scientific™ QSP™ SAVIT white elastomer co-polymer 12/13mm cap fits inside the rim 12mm and 13mm tubes and over the rim of 10mm tubes. Non-autoclavable. Packaged in bulk bags and non-sterile clean claims are; RNase/DNase, DNA, pyrogen and endotoxin free.
  • Polyethylene material is non autoclavable
  • Variety of colors
  • 사양
    클린 클레임RNase/DNase, DNA, pyrogen and endotoxin free
    마개 크기16 mm
    오토클레이브 가능Not Autoclavable
    색상White
    물질Elastomer Co-polymer
    멸균Non-sterile
    Unit SizeCase of 10000