Avidin, Alexa Fluor™ 488 conjugate
Invitrogen™

Avidin, Alexa Fluor™ 488 conjugate

Avidin, Alexa Fluor™ 488 conjugate
Have Questions?
產品號碼Quantity
A213701 mg
產品號碼 A21370
價格 (TWD)
9,440.00
線上優惠
Ends: 31-Dec-2026
11,800.00
您節省 2,360.00 (20%)
Each
Quantity:
1 mg
Request bulk or custom format
價格 (TWD)
9,440.00
線上優惠
Ends: 31-Dec-2026
11,800.00
您節省 2,360.00 (20%)
Each

Our avidin, Alexa Fluor™ 488 conjugate is a bright, photostable probe with excitation and emission characteristics similar to those of fluorescein. Avidin is a glycosylated biotin-binding protein that has a basic isoelectric point and is widely used to detect biotinylated probes.

For Research Use Only. Not for use in diagnostic procedures.

規格
Label or DyeAlexa Fluor Dyes
Quantity1 mg
Shipping ConditionRoom Temperature
ConjugateAlexa Fluor 488
FormSolid
Product LineAlexa Fluor
Unit SizeEach
內容物與存放
Store in freezer (-5 to -30°C) and protect from light.

引用資料與參考文獻 (20)

引用資料與參考文獻
Abstract
Association of excitatory amino acid transporters, especially EAAT2, with cholesterol-rich lipid raft microdomains: importance for excitatory amino acid transporter localization and function.
Authors:Butchbach ME, Tian G, Guo H, Lin CL
Journal:J Biol Chem
PubMed ID:15187084
In the present study, we investigated the role of membrane cholesterol in the function of glutamate transporters. Depletion of membrane cholesterol by methyl-beta-cyclodextrin resulted in reduced Na(+)-dependent glutamate uptake in primary cortical cultures. Glial glutamate transporter EAAT2-mediated uptake was more sensitive to this effect. Cell surface biotinylation and immunostaining experiments ... More
Identification of a subpopulation of cells with cancer stem cell properties in head and neck squamous cell carcinoma.
Authors:Prince ME, Sivanandan R, Kaczorowski A, Wolf GT, Kaplan MJ, Dalerba P, Weissman IL, Clarke MF, Ailles LE
Journal:Proc Natl Acad Sci U S A
PubMed ID:17210912
'Like many epithelial tumors, head and neck squamous cell carcinoma (HNSCC) contains a heterogeneous population of cancer cells. We developed an immunodeficient mouse model to test the tumorigenic potential of different populations of cancer cells derived from primary, unmanipulated human HNSCC samples. We show that a minority population of CD44(+) ... More
Prototype protein assembly as scaffold for time-resolved fluoroimmuno assays.
Authors:Barnhill HN, Claudel-Gillet S, Ziessel R, Charbonnière LJ, Wang Q
Journal:J Am Chem Soc
PubMed ID:17542581
'Turnip yellow mosaic virus (TYMV) is an icosahedral plant virus with an average diameter of 28 nm and can be isolated in gram quantities from turnip or Chinese cabbage inexpensively. In this study, it was selected as a prototype bionanoparticle for time-resolved fluoroimmuno assay (TRFIA). Two types of reactive amino ... More
Protein cargo delivery properties of cell-penetrating peptides. A comparative study.
Authors:Säälik P, Elmquist A, Hansen M, Padari K, Saar K, Viht K, Langel U, Pooga M
Journal:Bioconjug Chem
PubMed ID:15546190
'Application of cell-penetrating peptides for delivering various hydrophilic macromolecules with biological function into cells has gained much attention in recent years. We compared the protein transduction efficiency of four cell-penetrating peptides: penetratin, Tat peptide, transportan, and pVEC and studied the effects of various medium parameters on the uptake. Depletion of ... More
The role of alpha and beta chains in ligand recognition by beta 7 integrins.
Authors:Higgins JM, Cernadas M, Tan K, Irie A, Wang J, Takada Y, Brenner MB
Journal:J Biol Chem
PubMed ID:10837471
'Integrins alpha(E)beta(7) and alpha(4)beta(7) are involved in localization of leukocytes at mucosal sites. Although both alpha(E)beta(7) and alpha(4)beta(7) utilize the beta(7) chain, they have distinct binding specificities for E-cadherin and mucosal addressin cell adhesion molecule-1 (MAdCAM-1), respectively. We found that mutation of the metal ion-dependent adhesion site (MIDAS) in the ... More